missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SIK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29899
This item is not returnable.
View return policy
Description
SIK3 Polyclonal antibody specifically detects SIK3 in Human samples. It is validated for Immunoprecipitation
Specifications
| SIK3 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
| between 350 and 400 | |
| 100 μg | |
| Protein Kinase | |
| 23387 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction