Learn More
Invitrogen™ SIGN-R4 Monoclonal Antibody (210006)
GREENER_CHOICE
Product Code. 30535009
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535009 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30535009 Supplier Invitrogen™ Supplier No. MA560999
Rat Monoclonal Antibody
Reconstitute at 0.5 mg/mL in sterile PBS.
TRUSTED_SUSTAINABILITY
Specifications
Antigen SIGN-R4
Applications Western Blot
Classification Monoclonal
Clone 210006
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Cd209e
Gene Accession No. Q91ZW7
Gene Alias CD209; CD209 antigen-like protein E; Cd209e; CD209e antigen; DC-SIGNR4; DC-SIGN-related protein 4; mSIGNR4; SIGNR4
Gene Symbols Cd209e
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SIGNR4/CD209e, Ser41-Trp208.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170780
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b