missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIGLEC7 Recombinant Mouse Monoclonal Antibody (K70050_11B4)

Product Code. 31935031
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31935031 100 μL 100µL
31934617 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31935031 Supplier Invitrogen™ Supplier No. 780041M
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Recombinant mouse monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC7
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone K70050_11B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SIGLEC7
Gene Accession No. Q9Y286
Gene Alias Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2
Gene Symbols SIGLEC7
Host Species Mouse
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.