missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-3/CD33 Antibody (HIM3-4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
578.00€
Specifications
| Antigen | Siglec-3/CD33 |
|---|---|
| Clone | HIM3-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | HRP |
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human, Primate samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Siglec-3/CD33 | |
| Flow Cytometry | |
| HRP | |
| Mouse | |
| Immunology | |
| PBS | |
| 945 | |
| IgG1 | |
| Protein A purified |
| HIM3-4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
| NFMY-9s human cell line | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title