missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Siglec-10 Antibody (SIGLEC10/7582) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27039JF549
This item is not returnable.
View return policy
Description
Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Siglec-10 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7 | |
| Recombinant fragment (around aa500-697) of human Siglec-10 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 89790 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| SIGLEC10/7582 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction