missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SIB 1508Y maleate

Product Code. 18725047
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725047 10 mg 10mg
18776569 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18725047 Supplier Tocris Bioscience™ Supplier No. 4766/10
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent agonist of α4β2, α2β4, α4β4 and α3β4 nACh receptors

SIB 1508Y maleate is a potent neuronal nicotinic ACh receptor agonist (EC50 values are 1.8, 5, 9 and 23 nM for α4β2, α2β4, α4β4 and α3β4 receptors respectively). Improves cognitive function in a MPTP-induced model of Parkinson's Disease in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type SIB 1508Y maleate
Molecular Weight (g/mol) 302.33
Inhibitors SIB 1508Y maleate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.