missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shugoshin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20355
This item is not returnable.
View return policy
Description
Shugoshin Polyclonal specifically detects Shugoshin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Shugoshin | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| SGO1 | |
| Recombinant protein encompassing a sequence within the center region of human SGOL1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| FLJ14230, hSgo1, NY-BR-85, Serologically defined breast cancer antigen NY-BR-85, SGO, SGO1, shugoshin 1AB protein, shugoshin 1CD protein, shugoshin 1EF protein, shugoshin 1GH protein, shugoshin 1KL protein, shugoshin-like 1, shugoshin-like 1 (S. pombe) | |
| Rabbit | |
| 64 kDa | |
| 0.1 mL | |
| Cancer | |
| 151648 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction