missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Shiga toxin 2B Recombinant Rabbit Monoclonal Antibody (028)

Rabbit Recombinant Monoclonal Antibody
159.00€ - 429.00€
Specifications
| Antigen | Shiga toxin 2B |
|---|---|
| Clone | 028 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534836
![]() |
Invitrogen™
MA560353 |
100 μL |
429.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535145
|
Invitrogen™
MA560354 |
20 μL |
159.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Shiga toxin 2B Recombinant Monoclonal Antibody for ELISA
Shiga-like toxin (verotoxin) is a toxin produced by some strains of Escherichia coli. Shiga-like toxin is named for its similarity to the AB5-type Shiga toxin produced by the bacteria Shigella dysenteriae. There are two known types-SLT1 and SLT2. The Shiga-like toxin is linked with hemolytic-uremic syndrome. Shiga-like toxin requires highly specific receptors on the cells' surface in order to attach and enter the cell. Species such as cattle, swine, and deer which do not carry these receptors may harbor toxigenic bacteria without any ill effect, dropping them in their feces, from where they may be distributed to humans. Shiga Like Toxin-2 Subunit B has nontoxic action, it is the functional region which binds to the receptor. Upon binding to the host cell receptor, the Shiga Like Toxin-2 Subunit B forms a pentamer. The Shiga Like Toxin-2 Subunit B can be useful in vaccine study, antibody test and other functional research.Specifications
| Shiga toxin 2B | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Recombinant Enterohemorrhagic E. coli (EHEC) stx2B protein (Met1-Asp89). | |
| Antibody |
| 028 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Bacteria | |
| Shiga toxin; Shigatoxin Type 2; Shigatoxin Type II; STX2; STX-2b; STX-IIb | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title