Learn More
Sheep IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™
Product Code. 18106031
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18106031 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18106031 Supplier R&D Systems Supplier No. HAF016
  • ONLINE EXCLUSIVE PRICE
    237.00€
    223.65€ / 1mL
    Save 13.35€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Donkey Polyclonal Antibody has been used in 28 publications
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry∣Simple Western∣Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG
Applications Immunohistochemistry, Simple Western, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Immunogen Sheep IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects sheep IgG heavy and light chains in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with mouse IgG, rabbit IgG, human IgG, and chicken IgY is observed.
Target Species Sheep
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG