missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep IgG (H+L) PE-conjugated Antibody, R&D Systems™

Donkey Polyclonal Antibody
Brand: R&D Systems F0126
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry.
Specifications
| IgG | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| IGHG1 | |
| Sheep IgG | |
| 100 Tests | |
| Secondary | |
| Sheep IgG specific IgG was purified by sheep IgG (H+L) affinity chromatography and absorbed against human IgG, and human, mouse, bovine, and rabbit serum to eliminate cross-reactivity. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| R-PE | |
| Immunoglobulin G, ImmunoglobulinG | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction