missing translation for 'onlineSavingsMsg'
Learn More

Sheep IgG (H+L) APC-conjugated Antibody, R&D Systems™

Product Code. 18164051
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18164051 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18164051 Supplier R&D Systems Supplier No. F0127
  • ONLINE EXCLUSIVE PRICE
    408.00€
    385.35€ / 100 tests
    Save 22.65€
    5% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Immunogen Sheep IgG
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Sheep IgG specific IgG was purified by sheep IgG (H+L) affinity chromatography and absorbed against human IgG, and human, mouse, bovine, and rabbit serum to eliminate cross-reactivity.
Target Species Sheep
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.