missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Rat IgG (H+L) Secondary Antibody, PE/Atto594, Novus Biologicals™

Product Code. 18574891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18574891 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18574891 Supplier Novus Biologicals Supplier No. NB1206847PEATT594
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate PE/ATTO 594
Formulation PBS
Host Species Sheep
Immunogen Rat IgG whole molecule
Purification Method Multi-step
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.