missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Product Code. 18241900 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18241900 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18241900 Supplier Novus Biologicals Supplier No. NB1206796

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:500 - 1:2000, ELISA 1:15000, Immunohistochemistry 1:200 - 1:1000, Dot Blot
Gene Symbols IGHG
Host Species Sheep
Immunogen Rabbit IgG whole molecule
Purification Method Multi-step
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Alkaline Phosphatase (calf intestine), anti-Sheep Serum, Rabbit IgG and Rabbit Serum.
Target Species Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.