missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Mouse IgG2a Secondary Antibody, R&D Systems™

Product Code. 18634157 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18634157 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18634157 Supplier Novus Biologicals Supplier No. NBP269569
  • ONLINE EXCLUSIVE PRICE
    1023.11€
    930.30€ / 10mg
    Save 92.81€
    9% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a
Applications Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Formulation Lyophilized from PBS (pH 7.2)
Host Species Sheep
Immunogen Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.