Learn More
Sheep anti-Mouse IgG2a Secondary Antibody, Novus Biologicals™
Product Code. 18602817 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602817 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18602817 Supplier Novus Biologicals Supplier No. NBP269579
  • ONLINE EXCLUSIVE PRICE
    481.00€
    454.65€ / 1mL
    Save 26.35€
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
Sheep Polyclonal Antibody
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2a
Applications Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Whole antiserum
Host Species Sheep
Immunogen Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized