Learn More
Sheep anti-Mouse IgG2a Secondary Antibody, Biotin, Novus Biologicals™
Product Code. 18687376 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18687376 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18687376 Supplier Novus Biologicals Supplier No. NBP269570
Sheep Polyclonal Antibody
IgG2a Polyclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2a
Applications Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Formulation Lyophilized from PBS (pH 7.2)
Host Species Sheep
Immunogen Highly purified normal homogenous IgG2a(1b) isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Lyophilized