missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Mouse IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™

Product Code. 16273070
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16273070 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16273070 Supplier Abnova Supplier No. PAB10782.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Antigen Sheep anti-Mouse IgG whole molecule
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Sheep anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) gentamicLyophilized from sulfate (Do NOT add sodium azide!), 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free))
Host Species Sheep
Immunogen Mouse IgG whole molecule
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.