missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Human Lambda Light Chain Secondary Antibody, DyLight 550, Novus Biologicals™

Product Code. 30232241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30232241 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30232241 Supplier Novus Biologicals Supplier No. NB7482R
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Lambda Light Chain Polyclonal specifically detects Lambda Light Chain in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda Light Chain
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias IGLC, immunoglobulin lambda constant group
Host Species Sheep
Immunogen Human Lambda Free Light Chain
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3537
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.