missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human IgG (H+L) Secondary Antibody, PE/Cy5.5, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Dot Blot |
| Classification | Polyclonal |
| Conjugate | PE-Cyanine5.5 |
| Formulation | PBS |
| Host Species | Sheep |
| Immunogen | Human IgG whole molecule |
| Purification Method | Multi-step |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?