missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | TRITC |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen |
| Formulation | Lyophilized from PBS (pH 7.2) |
| Host Species | Sheep |
| Immunogen | Purified IgG prepared from pooled human serum. Freund's complete adjuvant is used in the first step of the immunization procedure. |
| Purification Method | Ion exchange chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?