missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Equine IgM Heavy Chain Secondary Antibody, Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Equine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Host Species | Sheep |
| Immunogen | Horse IgM |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?