missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Equine IgG (H+L) Secondary Antibody [Janelia Fluor 669], Novus Biologicals™

Product Code. 31851270 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31851270 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31851270 Supplier Novus Biologicals Supplier No. NB7433JF669
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Sheep
Immunogen This Sheep anti-Equine IgG (H+L) Secondary Antibody was developed against equine IgG-heavy and light chain.
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Equine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.