missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Equine IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB7433UV
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| This Sheep anti-Equine IgG (H+L) Secondary Antibody was developed against equine IgG-heavy and light chain. | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction