missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Canine IgG2 Secondary Antibody [Janelia Fluor 525], Novus Biologicals™

Product Code. 31882817 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31882817 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31882817 Supplier Novus Biologicals Supplier No. NBP268359JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG2 Polyclonal antibody specifically detects IgG2 in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Host Species Sheep
Immunogen Canine IgG2
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3501
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.