missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgM Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB761
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| HRP | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Bovine IgM | |
| 1 mL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this reacts specifically with bovine IgM. This may cross react with IgM from other species. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot Colorimetric 1:1000-1:10000 ECL 1:1000-1:30000, ELISA 1:10000-1:100000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Bovine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction