missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Bovine IgM Heavy Chain Secondary Antibody, DyLight 594, Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals NB758DL594

Product Code. 30226949

  • 541.00€ / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgM Heavy Chain
Polyclonal
50mM Sodium Borate
Sheep
Immunogen affinity purified
RUO
Secondary
Bovine
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 594
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Bovine IgM
1 mL
B Cell Development and Differentiation Markers, Immunology
3507
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.