Learn More
Sheep anti-Bovine IgM (H+L) Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-68322
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Immunodiffusion
Specifications
| IgM (H+L) | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Purified bovine IgM | |
| 1 mg | |
| B Cell Development and Differentiation Markers, Immunology | |
| Bovine | |
| Purified |
| ELISA, Immunodiffusion | |
| 1 mg/mL | |
| ELISA 1:100 - 1:1000, Immunodiffusion | |
| Sheep | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |