missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgM (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgM (H+L) Polyclonal specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Immunodiffusion.
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | ELISA, Immunodiffusion |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Host Species | Sheep |
| Immunogen | Purified bovine IgM |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?