missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Bovine IgM (H+L) Secondary Antibody, DyLight 488, Novus Biologicals™

Product Code. 30227042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30227042 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30227042 Supplier Novus Biologicals Supplier No. NBP268322G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgM (H+L) Polyclonal specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Immunodiffusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM (H+L)
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Host Species Sheep
Immunogen Purified bovine IgM
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.