missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG1 Secondary Antibody, Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG1 Polyclonal specifically detects IgG1 in Bovine samples. It is validated for ELISA, Immunodiffusion.
Specifications
Specifications
| Antigen | IgG1 |
| Applications | ELISA, Immunodiffusion |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | IgG1, Igh-4, RGD1359539, VH7183 |
| Host Species | Sheep |
| Immunogen | Purified bovine IgG1 |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?