missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgA Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgA Polyclonal specifically detects IgA in Bovine samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.
Specifications
Specifications
| Antigen | IgA |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Immunoglobulin A |
| Host Species | Sheep |
| Immunogen | Purified bovine IgA |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?