missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Bovine IgA Secondary Antibody, Biotin, Novus Biologicals™

Product Code. 30229411 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30229411 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30229411 Supplier Novus Biologicals Supplier No. NBP268347B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgA Polyclonal antibody specifically detects IgA in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Immunoglobulin A
Host Species Sheep
Immunogen Purified bovine IgA
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.