missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Bovine IgA Heavy Chain Secondary Antibody, FITC, Novus Biologicals™

Product Code. 18134319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
FITC
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate unitSize
18134319 1 mL FITC 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18134319 Supplier Novus Biologicals Supplier No. NBP246601F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Specifically detects IgA in Bovine samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry

IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA Heavy Chain
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500
Formulation PBS with 0.09% Sodium Azide
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Sheep
Immunogen Bovine IgA
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with bovine IgA This antibody may cross react with IgA from other species.
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.