Description
Demonstrate polysaccharides, neutral mucosubstances and basement membranes with the Thermo Scientific™ Shandon™ Periodic Acid Schiff Kit (PAS).
Includes:
Digestion kit, Harris Acidified Hematoxylin counterstain, manual, instrument and microwave protocols
Specifications
Specifications
| Type | Periodic Acid Schiff |
| Quantity | 250 mL |