Learn More
Shandon™ Periodic Acid Schiff Kit (PAS)
Product Code. 10455129 Shop All Epredia Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10455129 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10455129 Supplier Epredia Supplier No. B24200250
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Demonstrate polysaccharides, neutral mucosubstances and basement membranes with the Thermo Scientific™ Shandon™ Periodic Acid Schiff Kit (PAS)
Demonstrate polysaccharides, neutral mucosubstances and basement membranes with the Thermo Scientific™ Shandon™ Periodic Acid Schiff Kit (PAS).
Includes:
Digestion kit, Harris Acidified Hematoxylin counterstain, manual, instrument and microwave protocols
TRUSTED_SUSTAINABILITY
Specifications
Type Periodic Acid Schiff
Quantity 250 mL