missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3RF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76284-0.025mg
This item is not returnable.
View return policy
Description
SH3RF2 Polyclonal specifically detects SH3RF2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SH3RF2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, Heart protein phosphatase 1-binding protein, HEPP1FLJ23654, MGC149788, MGC149789, putative E3 ubiquitin-protein ligase SH3RF2, RING finger protein 158, RNF158MGC90410, SH3 domain containing ring finger 2, SH3 domain-containing RING finger protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 20 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_689763 | |
| SH3RF2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SH3RF2. The immunogen is located within the last 50 amino acids of SH3RF2. Amino Acid Squence: CHSGQQTDLRRKSALGK | |
| 0.025 mg | |
| Zinc Finger | |
| 153769 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction