missing translation for 'onlineSavingsMsg'
Learn More

SH3BGRL Monoclonal Antibody (OTI1E5), TrueMAB™, OriGene

Product Code. 17562234
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17562234 30 μL 30µL
17542234 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17562234 Supplier Origene Technologies Supplier No. TA809716S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human samples. It is validated for Western Blot

Specifications

Antigen SH3BGRL
Applications Western Blot
Classification Monoclonal
Clone OTI1E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDCL3
Gene Accession No. Q9H2J4
Gene Alias 1190008F14Rik; epididymis secretory protein Li 115; HEL-S-115; RGD1560520; SH3 domain binding glutamate rich protein like; SH3 domain binding glutamate-rich protein like; SH3 domain binding glutamic acid-rich protein like; SH3 domain-binding glutamic acid-rich-like protein; SH3BGR; Sh3bgrl; SH3-binding domain glutamic acid-rich protein like; similar to SH3 domain binding glutamic acid-rich protein like; zgc:64130; zSH3BGRL
Host Species Mouse
Immunogen Full length human recombinant protein of human PDCL3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79031
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.