missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BGRL Antibody (OTI8F7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45012-30ul
This item is not returnable.
View return policy
Description
SH3BGRL Monoclonal antibody specifically detects SH3BGRL in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| SH3BGRL | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI8F7 | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| MGC117402, SH3 domain binding glutamic acid-rich protein like, SH3 domain-binding glutamic acid-rich-like protein, SH3BGR, SH3-binding domain glutamic acid-rich protein like | |
| Full length human recombinant protein of human SH3BGRL (NP_003013) produced in E.coli. | |
| 30 μL | |
| Signal Transduction | |
| 6451 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction