missing translation for 'onlineSavingsMsg'
Learn More

SH2B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

195.00€

Specifications

Antigen SH2B2
Dilution Western Blot 1:500
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18763014
View Documents
Novus Biologicals
NBP2-22515-0.025mg
0.025 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

SH2B2 Polyclonal specifically detects SH2B2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

SH2B2
Western Blot
Unconjugated
RUO
PBS with 1 mg/mL BSA with 0.05% Sodium Azide
10603
Synthetic peptide corresponding to residues E(605) A T L G R A R A V E N Q Y S(619) from mouse APS.
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500
Polyclonal
Rabbit
Mouse
adaptor protein with pleckstrin homology and src homology 2 domains, APSAdapter protein with pleckstrin homology and Src homology 2 domains, SH2 and PH domain-containing adapter protein APS, SH2B adapter protein 2, SH2B adaptor protein 2
SH2B2
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.