Learn More
Trajan™ Thermo Scientific Instrument Ferrules
Product Code. 10493392
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
0.32 mm
0.45 to 0.53 mm
Unit Size:
1 set
Each
Product Code. 10493392
Brand: Trajan™ 073451
Only null left
Add to basket
Add to basket
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type GC Interface Connection
Material SilTite Metal
Quantity 10 ferrules, 2 nuts
Diameter (Metric) Inside 0.32 mm