Learn More
SGE™ GC Septa: CS
Product Code. p-3244804
Click to view available options
Diameter (English) Inner:
0.18 in.
0.37 in.
0.39 in.
0.43 in.
0.45 in.
0.5 in.
0.59 in.
Unit Size:
Pack of 48
Pack of 50
Product Code. 10095260
Brand: SGE™ 041825
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed specifically for use in Agilent Technologies™ (HP) GC instruments
CS Septa
  • PTFE-coated silicone septa for low temperature applications
  • Good durability, good resealing
  • Excellent solvent resistance; good tear resistance
  • Maximum temperature: 200°C (392°F)
Specifications
Diameter (English) Inner 0.39 in.
Material PTFE-coated Silicone
Diameter (Metric) Inner 10 mm
Quantity 50/Pk.