Learn More
Trajan™ EasyLOK Fittings
Product Code. 15298903
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 15298903
Brand: Trajan™ 0730014
Only null left
Add to basket
Add to basket
Ideal for applications where corrosive solvents are being used. Recommended for use with SGE GLT™ columns
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Type Replacement PEEK Hexnut Ferrule
Max. Pressure 5000 psi
Thread Size 10-32
Material Stainless Steel
Quantity 10/Pk.
Includes Pk. size: 10