missing translation for 'onlineSavingsMsg'
Learn More

SFRS12 Antibody, Novus Biologicals™

Product Code. 18159871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18159871 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18159871 Supplier Novus Biologicals Supplier No. NBP220332

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SFRS12 Polyclonal specifically detects SFRS12 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS12
Applications Western Blot
Classification Polyclonal
Concentration 0.75mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8WXA9
Gene Alias DKFZp564B176, Serine/arginine-rich-splicing regulatory protein 86, SFRS12serine-arginine-rich-splicing regulatory protein 86, Splicing factor, arginine/serine-rich 12MGC133045, splicing regulatory glutamine/lysine-rich protein 1, Splicing regulatory protein 508, SRRp508, SRrp508SRrp86serine-arginine-rich splicing regulatory protein 508, SRRP86
Gene Symbols SREK1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SFRS12. The exact sequence is proprietary.
Molecular Weight of Antigen 59 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140890
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.