missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38091-20ul
This item is not returnable.
View return policy
Description
SF3A1 Polyclonal antibody specifically detects SF3A1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SF3A1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| pre-mRNA processing 21, pre-mRNA splicing factor SF3a (120 kDa subunit), Prp21, PRPF21, SAP 114, SAP114splicing factor 3a, subunit 1, 120kD, SF3a120, spliceosome associated protein 114, Spliceosome-associated protein 114, splicing factor 3 subunit 1, splicing factor 3A subunit 1, splicing factor 3a, subunit 1, 120kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 715-785 of human SF3A1 (Q15459).,, Sequence:, QDKTEWKLNGQVLVFTLPLTDQVSVIKVKIHEATGMPAGKQKLQYEGIFIKDSNSLAYYNMANGAVIHLAL | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10291 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction