missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35675-100ul
This item is not returnable.
View return policy
Description
SF2 Polyclonal antibody specifically detects SF2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Alternative-splicing factor 1, ASFpre-mRNA-splicing factor SF2, P33 subunit, MGC5228, serine/arginine-rich splicing factor 1, SF2FLJ53078, SF2p33, SFRS1splicing factor 2, Splicing factor, arginine/serine-rich 1ASF-1, SR splicing factor 1, SRp30a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-248 of human SF2 (NP_008855.1).,, Sequence:, GGGGGGGAPRGRYGPPSRRSENRVVVSGLPPSGSWQDLKDHMREAGDVCYADVYRDGTGVVEFVRKEDMTYAVRKLDNTKFRSHEGETAYIRVKVDGPRSP | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 6426 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction