missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Antibody (CL9101) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88921AF700
This item is not returnable.
View return policy
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SF-1/NR5A1/Steroidogenic Factor 1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
| This antibody was developed using a synthetic peptide derived from Q13285, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology | |
| 2516 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL9101 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction