Learn More
Seven Components Solution for Ion Chromatography, H2O Matrix, Fisher Chemical™
Product Code. 12954305 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12954305 100 mL 100mL
12974435 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12954305 Supplier Fisher Chemical Supplier No. CM 1429
  • ONLINE EXCLUSIVE PRICE
    287.57€
    258.80€ / 100mL
    Save 28.77€
    10% Off
 Restricted Item
Estimated Shipment: 11-12-2026