missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETDB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20322
This item is not returnable.
View return policy
Description
SETDB1 Polyclonal antibody specifically detects SETDB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Proximity Ligation Assay, ChIP assay, Immunocytochemistry.
Specifications
| SETDB1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
| ERG-associated protein with SET domain, ESETEC 2.1.1.43, H3-K9-HMTase 4, Histone H3-K9 methyltransferase 4, histone-lysine N-methyltransferase SETDB1, histone-lysine N-methyltransferase, H3lysine-9 specific 4, KG1T, KIAA0067H3-K9-HMTase4, KMT1EERG-associated protein with a SET domain, ESET, Lysine N-methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1 | |
| Rabbit | |
| 143 kDa | |
| 0.1 mL | |
| Epigenetics, Transcription Factors and Regulators | |
| 9869 | |
| Human, Mouse | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay | |
| 0.63mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry | |
| Q15047 | |
| SETDB1 | |
| Recombinant protein encompassing a sequence within the center region of human SETDB1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (83%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction