Learn More
MP Biomedicals™ Set of 4 dNTPs (100mm)
Product Code. 11467110 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
4 x 100 μmol
4 x 25 μmol
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467110 4 x 100 μmol Pack of 4
11457110 4 x 25 μmol Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11467110 Supplier MP Biomedicals™ Supplier No. 11NTACG111
  • ONLINE EXCLUSIVE PRICE
    605.00€
    575.40€ / Pack of 4
    Save 29.60€
    4% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Offers maximum flexibility in preparation of reaction mixes. MP Biomedicals Set of 4 dNTPs (100mm) includes four separate vials of highly purified sodium salts of dATP, dCTP, dGTP and dTTP. These solutions achieve sensitive and consistent PCR results.
This product allows the addition of each dNTP to be altered as required for specific experiments, such as:
  • When adding a radioactive nucleotide for labeling by PCR
  • To increase the chance of non-specific mutations
  • In PCR, real-time PCR, cDNA synthesis and DNA sequencing
TRUSTED_SUSTAINABILITY
Specifications
Concentration 100 mM
Content And Storage Set 4 DNTPs composed of four separate vials of sodium salts of dATP, dCTP, dGTP and dTTP. Store at -20°C.
Purity or Quality Grade 99+%
Quantity 4 x 100 μmol
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR