missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertraline, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Sertraline
Specifications
Specifications
| Chemical Name or Material | Sertraline |
| CAS | 79617-96-2 |
| Analyte or Component Names | Sertraline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 Cl2 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sertraline, 1-Naphthalenamine, 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-, (1S-cis)-, (+)-Sertraline, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthaleneamine, CP 51974 |
| InChI Formula | InChI=1S/C17H17Cl2N/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11/h2-6,8,10,12,17,20H,7,9H2,1H3/t12-,17-/m0/s1 |
| SMILES | CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| IUPAC Name | (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction