missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertraline Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Sertraline Hydrochloride |
| CAS | 79559-97-0 |
| Analyte or Component Names | Sertraline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 Cl2 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sertraline Hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride (1:1), (1S-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthaleneamine hydrochloride, (1S,4S)-N-Methyl-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, Altruline, Aremis, Atruline, Cp 51974-1, Dominum, Gladem, Jzoloft, Lesefer, Lustral, Selectra, Serlift, Sertraline hydrochloride, Seserine, Sirto, Sosser, Stimuloton, Zolof, Zoloft |
| InChI Formula | InChI=1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1 |
| Solution Type | Neat |
| SMILES | Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?